Merck
Simplified YM-26734 inhibitors of secreted phospholipase A2 group IIA.
Bioorganic & medicinal chemistry letters (2008-09-27)
Rob C Oslund, Nathan Cermak, Christophe L M J Verlinde, Michael H Gelb
ABSTRAKT
Simplified analogs of YM-26734, a known inhibitor of secreted phospholipase A(2) (sPLA(2)) group IIA, were synthesized and found to also display potent inhibition at low nanomolar concentrations. Analogs were based on the didodecanoylphloroglucinol portion of YM-26734 which contains the predicted active site calcium binding group.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
YM-26734, ≥95% (HPLC)
Informacje o cenach i dostępności nie są obecnie dostępne.