Merck
Correlation between reduction potentials and inhibitions of Epstein-Barr virus activation by anthraquinone derivatives.
Bioorganic & medicinal chemistry letters (2008-06-17)
Junko Koyama, Yu Nisino, Izumi Morita, Norihiro Kobayashi, Toshiyuki Osakai, Harukuni Tokuda
ABSTRAKT
As a continuation of studies using natural and synthetic products as cancer chemopreventive agents, we used cyclic voltammetry to examine the reduction-oxidation potentials of methylated emodin derivatives prepared from emodin in phosphate buffer at pH 7.2. A good correlation was found between the inhibitory effects on Epstein-Barr virus early antigen (EBV-EA) activation and the reduction potential of methylated emodin derivatives. Furthermore, there was significant correlation between EBV-EA activation and the reduction potential of 35 anthraquinone derivatives including methylated emodin derivatives. It was further shown that the correlation could be enhanced by including LUMO energy and the number of hydroxy groups as additional parameters.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anthraquinone, 97%
Supelco
Melting point standard 283-286°C, analytical standard
Sigma-Aldrich
Aloe-emodin, ≥95% (HPLC)
Sigma-Aldrich
Physcion, ≥98.0% (TLC)
Sigma-Aldrich
Emodin, from Frangula bark, ≥90% (HPLC)