Merck
Combined Proteomic and In Silico Target Identification Reveal a Role for 5-Lipoxygenase in Developmental Signaling Pathways.
Cell chemical biology (2018-09-27)
Silke Brand, Sayantani Roy, Peter Schröder, Bernd Rathmer, Jessica Roos, Shobhna Kapoor, Sumersing Patil, Claudia Pommerenke, Thorsten Maier, Petra Janning, Sonja Eberth, Dieter Steinhilber, Dennis Schade, Gisbert Schneider, Kamal Kumar, Slava Ziegler, Herbert Waldmann
ABSTRAKT
Identification and validation of the targets of bioactive small molecules identified in cell-based screening is challenging and often meets with failure, calling for the development of new methodology. We demonstrate that a combination of chemical proteomics with in silico target prediction employing the SPiDER method may provide efficient guidance for target candidate selection and prioritization for experimental in-depth evaluation. We identify 5-lipoxygenase (5-LO) as the target of the Wnt pathway inhibitor Lipoxygenin. Lipoxygenin is a non-redox 5-LO inhibitor, modulates the β-catenin-5-LO complex and induces reduction of both β-catenin and 5-LO levels in the nucleus. Lipoxygenin and the structurally unrelated 5-LO inhibitor CJ-13,610 promote cardiac differentiation of human induced pluripotent stem cells and inhibit Hedgehog, TGF-β, BMP, and Activin A signaling, suggesting an unexpected and yet unknown role of 5-LO in these developmental pathways.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Calcium Ionophore A23187, ≥98% (TLC), powder
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
2-Phospho-L-ascorbic acid trisodium salt, ≥95.0% (HPLC)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M1 antibody, clone M1, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Przeciwciało anty-β-katenina, Upstate®, from rabbit
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture