Merck
  • Oral and parenteral therapeutic options for outpatient urinary infections caused by enterobacteriaceae producing CTX-M extended-spectrum beta-lactamases.
Oral and parenteral therapeutic options for outpatient urinary infections caused by enterobacteriaceae producing CTX-M extended-spectrum beta-lactamases.
Antimicrobial agents and chemotherapy (2009-01-07)
Vidhya Prakash, James S Lewis, Monica L Herrera, Brian L Wickes, James H Jorgensen
ABSTRAKT
Effective therapeutic options are needed for community-onset urinary tract infections due to Escherichia coli strains that produce CTX-M extended-spectrum beta-lactamases. We examined 46 urinary isolates producing CTX-M against several oral or long-acting parenteral antimicrobial agents. Approximately 90% were susceptible to fosfomycin and to a combination of cefdinir plus amoxicillin-clavulanate. All were susceptible to ertapenem.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Nitrofurantoin, 98.0-102.0% (EP, UV), meets EP testing specifications
Supelco
Nitrofurantoin, VETRANAL®, analytical standard
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)