Merck
  • Anti-aging Effects of Mangosteen Peel Extract and Its Phytochemical Compounds: Antioxidant Activity, Enzyme Inhibition and Molecular Docking Simulation.
Anti-aging Effects of Mangosteen Peel Extract and Its Phytochemical Compounds: Antioxidant Activity, Enzyme Inhibition and Molecular Docking Simulation.
Tropical life sciences research (2020-11-21)
Wahyu Widowati, Chrismis Novalina Ginting, I Nyoman Ehrich Lister, Ermi Girsang, Annisa Amalia, Satrio Haryo Benowo Wibowo, Hanna Sari Widya Kusuma, Rizal
ABSTRAKT
Skin aging is a complex natural process characterised by gradual diminishment of structural integrity and physiological imbalance of the skin tissue. Since the oxidative stress is tightly corelated to the skin aging process, the usage of antioxidant may serve as favourable strategies for slowing down the skin aging process. Mangosteen is an important fruit commodity and its extract had been extensively studied and revealing various biological activities. Present study aimed to assess the antioxidant and antiaging activity of mangosteen peel extract (MPE) and its phytochemical compounds. MPE and its compounds were subjected to ferric reducing antioxidant power (FRAP), hydroperoxide (H2O2) scavenging, anti-collagenase, anti-elastase, anti-hyaluronidase and anti-tyrosinase assay. MPE has the highest FRAP 116.31 ± 0.60 μM Fe(II) μg-1 extract, IC50 of MPE on H2O2 scavenging activity was 54.61 μg mL-1. MPE also has the highest anti elastase activity at IC50 7.40 μg mL-1. Alpha-mangostin showed potent anti-collagenase activity (IC50 9.75 μg mL-1). While gamma-mangostin showed potent anti-hyaluronidase (IC50 23.85 μg mL-1) and anti-tyrosinase (IC50 50.35 μg mL-1). MPE and its compounds were evaluated in vitro for antioxidant and antiaging activities. Current findings may provide scientific evidence for possible usage of mangosteen extract and its compounds as antioxidant and antiaging agent.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Sigma Blend Type H, ≥1.0 FALGPA units/mg solid
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium acetate buffer solution, pH 5.2±0.1 (25 °C), Molecular Biology, 3 M, 0.2 μm filtered
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
N-[3-(2-Furyl)acryloyl]-Leu-Gly-Pro-Ala
Informacje o cenach i dostępności nie są obecnie dostępne.