Merck
Pyruvate affects inflammatory responses of macrophages during influenza A virus infection.
Virus research (2020-07-08)
Hazar Abusalamah, Jessica M Reel, Christopher R Lupfer
ABSTRAKT
Pyruvate is the end product of glycolysis and transported into the mitochondria for use in the tricarboxylic acid (TCA) cycle. It is also a common additive in cell culture media. We discovered that inclusion of sodium pyruvate in culture media during infection of mouse bone marrow derived macrophages with influenza A virus impaired cytokine production (IL-6, IL-1β, and TNF-α). Sodium pyruvate did not inhibit viral RNA replication. Instead, the addition of sodium pyruvate alters cellular metabolism and diminished mitochondrial reactive oxygen species (ROS) production and lowered immune signaling. Overall, sodium pyruvate affects the immune response produced by macrophages but does not inhibit virus replication.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose and sodium bicarbonate, without L-glutamine and sodium pyruvate, liquid, sterile-filtered, suitable for cell culture, suitable for hybridoma
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O127:B8, purified by phenol extraction
Informacje o cenach i dostępności nie są obecnie dostępne.