Merck
Effects of 7-O substitutions on estrogenic and anti-estrogenic activities of daidzein analogues in MCF-7 breast cancer cells.
Journal of medicinal chemistry (2010-07-31)
Quan Jiang, Florastina Payton-Stewart, Steven Elliott, Jennifer Driver, Lyndsay V Rhodes, Qiang Zhang, Shilong Zheng, Deepak Bhatnagar, Stephen M Boue, Bridgette M Collins-Burow, Jayalakshmi Sridhar, Cheryl Stevens, John A McLachlan, Thomas E Wiese, Matthew E Burow, Guangdi Wang
ABSTRAKT
Daidzein (1) is a natural estrogenic isoflavone. We report here that 1 can be transformed into anti-estrogenic ligands by simple alkyl substitutions of the 7-hydroxyl hydrogen. To test the effect of such structural modifications on the hormonal activities of the resulting compounds, a series of daidzein analogues have been designed and synthesized. When MCF-7 cells were treated with the analogues, those resulting from hydrogen substitution by isopropyl (3d), isobutyl (3f), cyclopentyl (3g), and pyrano- (2) inhibited cell proliferation, estrogen-induced transcriptional activity, and estrogen receptor (ER) regulated progesterone receptor (PgR) gene expression. However, methyl (3a) and ethyl (3b) substitutions of the hydroxyl proton only led to moderate reduction of the estrogenic activities. These results demonstrated the structural requirements for the transformation of daidzein from an ER agonist to an antagonist. The most effective analogue, 2, was found to reduce in vivo estrogen stimulated MCF-7 cell tumorigenesis using a xenograft mouse model.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Daidzein, ≥98%, synthetic
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
Estradiol, meets USP testing specifications