Merck
The glucocorticoid receptor recruits the COMPASS complex to regulate inflammatory transcription at macrophage enhancers.
Cell reports (2021-02-11)
Franziska Greulich, Michael Wierer, Aikaterini Mechtidou, Omar Gonzalez-Garcia, N Henriette Uhlenhaut
ABSTRAKT
Glucocorticoids (GCs) are effective anti-inflammatory drugs; yet, their mechanisms of action are poorly understood. GCs bind to the glucocorticoid receptor (GR), a ligand-gated transcription factor controlling gene expression in numerous cell types. Here, we characterize GR's protein interactome and find the SETD1A (SET domain containing 1A)/COMPASS (complex of proteins associated with Set1) histone H3 lysine 4 (H3K4) methyltransferase complex highly enriched in activated mouse macrophages. We show that SETD1A/COMPASS is recruited by GR to specific cis-regulatory elements, coinciding with H3K4 methylation dynamics at subsets of sites, upon treatment with lipopolysaccharide (LPS) and GCs. By chromatin immunoprecipitation sequencing (ChIP-seq) and RNA-seq, we identify subsets of GR target loci that display SETD1A occupancy, H3K4 mono-, di-, or tri-methylation patterns, and transcriptional changes. However, our data on methylation status and COMPASS recruitment suggest that SETD1A has additional transcriptional functions. Setd1a loss-of-function studies reveal that SETD1A/COMPASS is required for GR-controlled transcription of subsets of macrophage target genes. We demonstrate that the SETD1A/COMPASS complex cooperates with GR to mediate anti-inflammatory effects.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Fetal Bovine Serum, Heat Inactivated, non-USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Roche
cOmplete, Mini, Koktajl inhibitorów proteaz bez EDTA, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Octoclothepin maleate salt, solid
Sigma-Aldrich
Przeciwciało anty-trimetylo-histonowe H3 (Lys4), klon 15-10C-E4, królicze monoklonalne, clone 15-10C-E4, Upstate®, from rabbit