Merck
Phytochemical and Psychotropic Research of Motherwort (Leonurus cardiaca L.) Modified Dry Extracts.
Plants (Basel, Switzerland) (2021-01-29)
Oleh Koshovyi, Ain Raal, Igor Kireyev, Nadiya Tryshchuk, Tetiana Ilina, Yevhen Romanenko, Sergiy M Kovalenko, Natalya Bunyatyan
ABSTRAKT
The prospect of creating a new medicine with psychotropic activity is shown as a result of studying the chemical composition and pharmacological activity of modified dry extracts of motherwort (Leonurus cardiaca L.) tincture. The most promising substances were the dry extracts, modified by adding small amounts of arginine, valine, phenylalanine, glycine, lysine, and alanine. A total of 15 main phenolic substances were found in the extracts, and eight of them were identified. There were also 10 hydroxycinnamic acids in these extracts, three of which were identified (chlorogenic, caffeic, and rosmarinic acids). The dominant hydroxycinnamic acids were chlorogenic and caffeic acids. Among flavonoids, catechin, hyperoside, and rutin were identified. It should be noted that the extracts had a significant content of ellagic acid. On the basis of the results of the phytochemical analysis of the extracts, it can be concluded that the composition of phenolic compounds does not differ significantly, and the main differences are related to amino acids, which obviously have an impact on the overall pharmacological effect. The results obtained indicate the presence of anxiolytic activity in the motherwort extracts studied in complex with amino acids. The extracts with glycine, valine, and arginine were more effective in reducing anxiety in animals.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Rosmarinic acid, ≥98% (HPLC), from Rosemarinus officinalis L.
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Sigma-Aldrich
Rutin hydrate, ≥94% (HPLC), powder
Sigma-Aldrich
Ellagic acid, ≥95% (HPLC), powder, from tree bark
Sigma-Aldrich
Hydroxylamine hydrochloride, ACS reagent, 98.0%
Millipore
ReBlot Plus Kit
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
Quercetin 3-D-galactoside, ≥97.0% (HPLC)