Merck
  • Use of Zebrafish Embryo Assay to Evaluate Toxicity and Safety of Bioreactor-Grown Exopolysaccharides and Endopolysaccharides from European Ganoderma applanatum Mycelium for Future Aquaculture Applications.
Use of Zebrafish Embryo Assay to Evaluate Toxicity and Safety of Bioreactor-Grown Exopolysaccharides and Endopolysaccharides from European Ganoderma applanatum Mycelium for Future Aquaculture Applications.
International journal of molecular sciences (2021-02-11)
Wan Abd Al Qadr Imad Wan-Mohtar, Zul Ilham, Adi Ainurzaman Jamaludin, Neil Rowan
ABSTRAKT
Natural mycelial exopolysaccharide (EPS) and endopolysaccharide (ENS) extracted from bioreactor-cultivated European Ganoderma applanatum mushrooms are of potential high commercial value for both food and adjacent biopharmaceutical industries. In order to evaluate their potential toxicity for aquaculture application, both EPS (0.01-10 mg/mL) and ENS (0.01-10 mg/mL) extracts were tested for Zebrafish Embryo Toxicity (ZFET); early development effects on Zebrafish Embryos (ZE) were also analyzed between 24 and 120 h post-fertilization (HPF). Both EPS and ENS are considered non-toxic with LC50 of 1.41 mg/mL and 0.87 mg/mL respectively. Both EPS and ENS did not delay hatching and teratogenic defect towards ZE with <1.0 mg/mL, respectively. No significant changes in the ZE heart rate were detected following treatment with the two compounds tested (EPS: 0.01-10 mg/mL: 176.44 ± 0.77 beats/min and ENS: 0.01-10 mg/mL: 148.44 ± 17.75 beats/min) compared to normal ZE (120-180 beats/min). These initial findings support future pre-clinical trials in adult fish models with view to safely using EPS and ENS as potential feed supplements for supplements for development of the aquaculture industry.
MATERIAŁY
Numer produktu
Marka
Opis produktu
BRAND® 96-well microplate, U-bottom, round bottom, non-sterile
Informacje o cenach i dostępności nie są obecnie dostępne.