Merck
The secretome mouse provides a genetic platform to delineate tissue-specific in vivo secretion.
Proceedings of the National Academy of Sciences of the United States of America (2021-01-13)
Jie Liu, Ji Yong Jang, Mehdi Pirooznia, Shihui Liu, Toren Finkel
ABSTRAKT
At present, it remains difficult to deconvolute serum in order to identify the cell or tissue origin of a given circulating protein. Here, by exploiting the properties of proximity biotinylation, we describe a mouse model that enables the elucidation of the in vivo tissue-specific secretome. As an example, we demonstrate how we can readily identify in vivo endothelial-specific secretion as well as how this model allows for the characterization of muscle-derived serum proteins that either increase or decrease with exercise. This genetic platform should, therefore, be of wide utility in understanding normal and disease physiology and for the rational design of tissue-specific disease biomarkers.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Calnexin Antibody, from rabbit, purified by affinity chromatography
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Triton X-100, laboratory grade
Informacje o cenach i dostępności nie są obecnie dostępne.