Merck
Benzenepolycarboxylic acids with potential anti-hemorrhagic properties and structure-activity relationships.
Bioorganic & medicinal chemistry (2011-11-04)
Hnin Thanda Aung, Toshiaki Nikai, Masatake Niwa, Yoshiaki Takaya
ABSTRAKT
Previously, we reported the structural requirements of the cinnamic acid relatives for inhibition of snake venom hemorrhagic action. In the present study, we examined the effect of benzenepolycarboxylic acids and substituted benzoic acids against Protobothropsflavoviridis venom-induced hemorrhage. Pyromellitic acid (1,2,4,5-benzenetetracarboxylic acid) was found to be a potent inhibitor of hemorrhage, with an IC(50) value of 0.035 μM. In addition, most of the antihemorrhagic activity of compounds tested in this experiment showed good correlation to acidity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
trans,trans-Muconic acid, 95%
Sigma-Aldrich
p-Toluic acid, 98%
Sigma-Aldrich
2-Chlorobenzoic acid, 98%
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
Sigma-Aldrich
Trimesic acid, 95%
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Terephthalic acid, 98%
Sigma-Aldrich
Phthalic acid, puriss. p.a., ≥99.5% (T)
Sigma-Aldrich
Phthalic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Phthalic acid, reagent grade, 98%
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Mellitic acid, 99%
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
1,2,4-Benzenetricarboxylic acid, ≥99%
Sigma-Aldrich
1,2,4,5-Benzenetetracarboxylic acid, 96%
Sigma-Aldrich
4-Chlorobenzoic acid, 99%
Sigma-Aldrich
3-Hydroxybenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Supelco
Benzoic acid, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland