- Verrucamides A-D, antibacterial cyclopeptides from Myrothecium verrucaria.
Verrucamides A-D, antibacterial cyclopeptides from Myrothecium verrucaria.
Journal of natural products (2011-05-05)
Xianwei Zou, Shubin Niu, Jinwei Ren, Erwei Li, Xingzhong Liu, Yongsheng Che
PMID21539317
ABSTRAKT
Four new cyclic tetradecapeptides, verrucamides A-D (1-4), have been isolated from the solid-substrate fermentation culture of the ascomycete fungus Myrothecium verrucaria. The structures of these compounds, each featuring six N-methylated amino acid residues, were elucidated primarily by NMR and MS methods. The absolute configurations of 1-4 were assigned by application of Marfey's method on their acid hydrolysates. Compounds 1-4 showed antimicrobial activity against the Gram-positive bacterium Staphylococcus aureus.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Informacje o cenach i dostępności nie są obecnie dostępne.