Merck
In Vitro Derivation of Melanocytes from Embryonic Neural Crest Stem Cells.
Methods in molecular biology (Clifton, N.J.) (2015-12-15)
Olga Shakhova, Lukas Sommer
ABSTRAKT
During development, melanocyte progenitors originate from the neural crest, a transient embryonic structure in vertebrates that gives rise to a variety of cell types including neurons and glia of the peripheral nervous system, smooth muscle cells of the cardiovascular system, chondrocytes and osteoblasts of the craniofacial elements, and pigment cells in the skin. In this chapter, we describe a method for the differentiation of multipotent embryonic neural crest stem cells into differentiated pigmented melanocytes by using in vitro explant culture system. This protocol allows the dissection of genetic and cellular mechanisms regulating neural crest stem cell and melanocyte development. Based on this knowledge it is possible to make predictions about processes that might also be implicated in melanoma initiation and progression.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
α-Melanocyte stimulating hormone, ≥97% (HPLC)
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Endothelin 3 human, rat, ≥97% (HPLC), powder
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%