Merck
Nitrogenase Fe protein: A molybdate/homocitrate insertase.
Proceedings of the National Academy of Sciences of the United States of America (2006-10-26)
Yilin Hu, Mary C Corbett, Aaron W Fay, Jerome A Webber, Keith O Hodgson, Britt Hedman, Markus W Ribbe
ABSTRAKT
The Fe protein is indispensable for nitrogenase catalysis and biosynthesis. However, its function in iron-molybdenum cofactor (FeMoco) biosynthesis has not been clearly defined. Here we show that the Fe protein can act as a Mo/homocitrate insertase that mobilizes Mo/homocitrate for the maturation of FeMoco precursor on NifEN. Further, we establish that Mo/homocitrate mobilization by the Fe protein likely involves hydrolysis of MgATP and protein-protein interaction between the Fe protein and NifEN. Our findings not only clarify the role of the Fe protein in FeMoco assembly and assign another function to this multitask enzyme but also provide useful insights into a mechanism of metal trafficking required for the assembly of complex metalloproteins such as nitrogenase.