Merck
Formaldehyde Vapor Characteristics in Varied Decontamination Environments.
Applied biosafety : journal of the American Biological Safety Association (2021-05-22)
Young W Choi, Michelle M Sunderman, Martha W McCauley, William R Richter, Zachary J Willenberg, Joseph Wood, Shannon Serre, Leroy Mickelsen, Stuart Willison, Rich Rupert, Jorge G Muñiz Ortiz, Sara Casey, M Worth Calfee
ABSTRAKT
This effort investigated formaldehyde vapor characteristics under various environmental conditions by the analyses of air samples collected over a time-course. This knowledge will help responders achieve desired formaldehyde exposure parameters for decontamination of affected spaces after a biological contamination incident. Prescribed masses of paraformaldehyde and formalin were sublimated or evaporated, respectively, to generate formaldehyde vapor. Adsorbent cartridges were used to collect air samples from the test chamber at predetermined times. A validated method was used to extract the cartridges and analyze for formaldehyde via liquid chromatography. In addition, material demand for the formaldehyde was evaluated by inclusion of arrays of Plexiglas panels in the test chamber to determine the impact of varied surface areas within the test chamber. Temperature was controlled with a circulating water bath connected to a radiator and fan inside the chamber. Relative humidity was controlled with humidity fixed-point salt solutions and water vapor generated from evaporated water. Low temperature trials (approximately 10°C) resulted in decreased formaldehyde air concentrations throughout the 48-hour time-course when compared with formaldehyde concentrations in the ambient temperature trials (approximately 22°C). The addition of clear Plexiglas panels to increase the surface area of the test chamber interior resulted in appreciable decreases of formaldehyde air concentration when compared to an empty test chamber. This work has shown that environmental variables and surface-to-volume ratios in the decontaminated space may affect the availability of formaldehyde in the air and, therefore, may affect decontamination effectiveness.
MATERIAŁY
Numer produktu
Marka
Opis produktu
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Paraformaldehyde, prilled, 95%
Informacje o cenach i dostępności nie są obecnie dostępne.