Merck
  • Cooperativity mediated by rationally selected combinations of human monoclonal antibodies targeting the henipavirus receptor binding protein.
Cooperativity mediated by rationally selected combinations of human monoclonal antibodies targeting the henipavirus receptor binding protein.
Cell reports (2021-09-02)
Michael P Doyle, Nurgun Kose, Viktoriya Borisevich, Elad Binshtein, Moushimi Amaya, Marcus Nagel, Edward J Annand, Erica Armstrong, Robin Bombardi, Jinhui Dong, Kevin L Schey, Christopher C Broder, Larry Zeitlin, Erin A Kuang, Zachary A Bornholdt, Brandyn R West, Thomas W Geisbert, Robert W Cross, James E Crowe
ABSTRAKT
Hendra virus and Nipah virus (NiV), members of the Henipavirus (HNV) genus, are zoonotic paramyxoviruses known to cause severe disease across six mammalian orders, including humans. We isolated a panel of human monoclonal antibodies (mAbs) from the B cells of an individual with prior exposure to equine Hendra virus (HeV) vaccine, targeting distinct antigenic sites. The most potent class of cross-reactive antibodies achieves neutralization by blocking viral attachment to the host cell receptors ephrin-B2 and ephrin-B3, with a second class being enhanced by receptor binding. mAbs from both classes display synergistic activity in vitro. In a stringent hamster model of NiV Bangladesh (NiVB) infection, antibodies from both classes reduce morbidity and mortality and achieve synergistic protection in combination. These candidate mAbs might be suitable for use in a cocktail therapeutic approach to achieve synergistic potency and reduce the risk of virus escape.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ouabain octahydrate, ≥95% (HPLC), powder
Sigma-Aldrich
Histopaque®-1077, sterile-filtered, density: 1.077
Sigma-Aldrich
Cyclosporin A, BioReagent, from Tolypocladium inflatum, Molecular Biology, ≥95%
Sigma-Aldrich
Chk2 Inhibitor II hydrate, ≥98% (HPLC)
Sigma-Aldrich
Suplement pożywki HAT (50×) Hybri-Max, γ-irradiated, lyophilized powder, BioXtra, suitable for hybridoma
Sigma-Aldrich
HT Media Supplement (50×) Hybri-Max, lyophilized powder, γ-irradiated, BioXtra, suitable for hybridoma