Merck
  • Encapsulated Pine Bark Polyphenolic Extract during Gastrointestinal Digestion: Bioaccessibility, Bioactivity and Oxidative Stress Prevention.
Encapsulated Pine Bark Polyphenolic Extract during Gastrointestinal Digestion: Bioaccessibility, Bioactivity and Oxidative Stress Prevention.
Foods (Basel, Switzerland) (2021-02-10)
Pedro Ferreira-Santos, Raquel Ibarz, Jean-Michel Fernandes, Ana Cristina Pinheiro, Cláudia Botelho, Cristina M R Rocha, José António Teixeira, Olga Martín-Belloso
ABSTRAKT
Polyphenolic extracts from pine bark have reported different biological actions and promising beneficial effects on human health. However, its susceptibility to environmental stresses (temperature, storage, etc.) and physiological human conditions prequires the development of efficient protection mechanisms to allow effective delivering of functionality. The aim of this work was to encapsulate pine bark extract rich phenolic compounds by spray-drying using maltodextrin, and understand the influence of encapsulation on the antioxidant and antimicrobial activity and bioaccessibility of phenolic compounds during gastrointestinal digestion. The optimized process conditions allowed good encapsulation efficiency of antioxidant phenolic compounds. The microencapsulation was effective in protecting those compounds during gastrointestinal conditions, controlling their delivery and enhancing its health benefits, decreasing the production of reactive oxygen species implicated in the process of oxidative stress associated with some pathologies. Finally, this encapsulation system was able to protect these extracts against acidic matrices, making the system suitable for the nutritional enrichment of fermented foods or fruit-based beverages, providing them antimicrobial protection, because the encapsulated extract was effective against Listeria innocua. Overall, the designed system allowed protecting and appropriately delivering the active compounds, and may find potential application as a natural preservative and/or antioxidant in food formulations or as bioactive ingredient with controlled delivery in pharmaceuticals or nutraceuticals.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
α-Amylase from human saliva, Type XIII-A, lyophilized powder, 300-1,500 units/mg protein
Sigma-Aldrich
Pancreatin from porcine pancreas, 8 × USP specifications
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥2,500 units/mg protein (E1%/280)