Merck
dCas9 fusion to computer-designed PRC2 inhibitor reveals functional TATA box in distal promoter region.
Cell reports (2022-03-03)
Shiri Levy, Logeshwaran Somasundaram, Infencia Xavier Raj, Diego Ic-Mex, Ashish Phal, Sven Schmidt, Weng I Ng, Daniel Mar, Justin Decarreau, Nicholas Moss, Ammar Alghadeer, Henrik Honkanen, Jay Sarthy, Nicholas Vitanza, R David Hawkins, Julie Mathieu, Yuliang Wang, David Baker, Karol Bomsztyk, Hannele Ruohola-Baker
ABSTRAKT
Bifurcation of cellular fates, a critical process in development, requires histone 3 lysine 27 methylation (H3K27me3) marks propagated by the polycomb repressive complex 2 (PRC2). However, precise chromatin loci of functional H3K27me3 marks are not yet known. Here, we identify critical PRC2 functional sites at high resolution. We fused a computationally designed protein, EED binder (EB), which competes with EZH2 and thereby inhibits PRC2 function, to dCas9 (EBdCas9) to allow for PRC2 inhibition at a precise locus using gRNA. Targeting EBdCas9 to four different genes (TBX18, p16, CDX2, and GATA3) results in precise H3K27me3 and EZH2 reduction, gene activation, and functional outcomes in the cell cycle (p16) or trophoblast transdifferentiation (CDX2 and GATA3). In the case of TBX18, we identify a PRC2-controlled, functional TATA box >500 bp upstream of the TBX18 transcription start site (TSS) using EBdCas9. Deletion of this TATA box eliminates EBdCas9-dependent TATA binding protein (TBP) recruitment and transcriptional activation. EBdCas9 technology may provide a broadly applicable tool for epigenomic control of gene regulation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Cas9 Protein, from Streptococcus pyogenes, recombinant, expressed in E. coli, 1X NLS
Sigma-Aldrich
Przeciwciało anty-EED, from rabbit
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid