Merck
Conformation-specific Antibodies Targeting Aggregated Forms of α-synuclein Block the Propagation of Synucleinopathy.
Experimental neurobiology (2022-03-09)
Minsun Choi, Tae-Kyung Kim, Jinhyung Ahn, Jun Sung Lee, Byung Chul Jung, Sungwon An, Dongin Kim, Min Jae Lee, Inhee Mook-Jung, Sang Hoon Lee, Seung-Jae Lee
ABSTRAKT
Abnormal aggregation of α-synuclein is a key element in the pathogenesis of several neurodegenerative diseases, including Parkinson's disease (PD), dementia with Lewy bodies, and multiple system atrophy. α-synuclein aggregation spreads through various brain regions during the course of disease progression, a propagation that is thought to be mediated by the secretion and subsequent uptake of extracellular α-synuclein aggregates between neuronal cells. Thus, aggregated forms of this protein have emerged as promising targets for disease-modifying therapy for PD and related diseases. Here, we generated and characterized conformation-specific antibodies that preferentially recognize aggregated forms of α-synuclein. These antibodies promoted phagocytosis of extracellular α-synuclein aggregates by microglial cells and interfered with cell-to-cell propagation of α-synuclein. In an α-synuclein transgenic model, passive immunization with aggregate-specific antibodies significantly ameliorated pathological phenotypes, reducing α-synuclein aggregation, gliosis, inflammation, and neuronal loss. These results suggest that conformation-specific antibodies targeting α-synuclein aggregates are promising therapeutic agents for PD and related synucleinopathies.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
ExtrAvidin®−Peroxidase, buffered aqueous solution
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for use with bacterial cell extracts, lyophilized powder
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
3, 3′,5 ,5′-Tetramethylbenzidine Liquid Substrate, Supersensitive, for ELISA, ready to use solution