Merck
KSHV episome tethering sites on host chromosomes and regulation of latency-lytic switch by CHD4.
Cell reports (2022-05-12)
Ashish Kumar, Yuanzhi Lyu, Yuichi Yanagihashi, Chanikarn Chantarasrivong, Vladimir Majerciak, Michelle Salemi, Kang-Hsin Wang, Tomoki Inagaki, Frank Chuang, Ryan R Davis, Clifford G Tepper, Kazushi Nakano, Chie Izumiya, Michiko Shimoda, Ken-Ichi Nakajima, Alexander Merleev, Zhi-Ming Zheng, Mel Campbell, Yoshihiro Izumiya
ABSTRAKT
Kaposi sarcoma-associated herpesvirus (KSHV) establishes a latent infection in the cell nucleus, but where KSHV episomal genomes are tethered and the mechanisms underlying KSHV lytic reactivation are unclear. Here, we study the nuclear microenvironment of KSHV episomes and show that the KSHV latency-lytic replication switch is regulated via viral long non-coding (lnc)RNA-CHD4 (chromodomain helicase DNA binding protein 4) interaction. KSHV episomes localize with CHD4 and ADNP proteins, components of the cellular ChAHP complex. The CHD4 and ADNP proteins occupy the 5'-region of the highly inducible lncRNAs and terminal repeats of the KSHV genome together with latency-associated nuclear antigen (LANA). Viral lncRNA binding competes with CHD4 DNA binding, and KSHV reactivation sequesters CHD4 from the KSHV genome, which is also accompanied by detachment of KSHV episomes from host chromosome docking sites. We propose a model in which robust KSHV lncRNA expression determines the latency-lytic decision by regulating LANA/CHD4 binding to KSHV episomes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Spermidine, ≥99% (GC)
Sigma-Aldrich
Przeciwciało przeciw polimerazie RNA II, klon CTD4H8, clone CTD4H8, Upstate®, from mouse
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-HHV-8 LNA-1 Antibody, clone LN53, clone LN53, from rat
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution