Merck
Identification of Pax protein inhibitors that suppress target gene expression and cancer cell proliferation.
Cell chemical biology (2021-11-26)
Shayna T J Bradford, Edward Grimley, Ann M Laszczyk, Pil H Lee, Sanjeevkumar R Patel, Gregory R Dressler
ABSTRAKT
The Pax family of developmental control genes are frequently deregulated in human disease. In the kidney, Pax2 is expressed in developing nephrons but not in adult proximal and distal tubules, whereas polycystic kidney epithelia or renal cell carcinoma continues to express high levels. Pax2 reduction in mice or cell culture can slow proliferation of cystic epithelial cells or renal cancer cells. Thus, inhibition of Pax activity may be a viable, cell-type-specific therapy. We designed an unbiased, cell-based, high-throughput screen that identified triazolo pyrimidine derivatives that attenuate Pax transactivation ability. We show that BG-1 inhibits Pax2-positive cancer cell growth and target gene expression but has little effect on Pax2-negative cells. Chromatin immunoprecipitation suggests that these inhibitors prevent Pax protein interactions with the histone H3K4 methylation complex at Pax target genes in renal cells. Thus, these compounds may provide structural scaffolds for kidney-specific inhibitors with therapeutic potential.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
BG-3, ≥98% (HPLC)
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Przeciwciało anty-fosfo-histonowe H3 (Ser10), marker mitozy, Upstate®, from rabbit