Merck
  • Poly(I:C) promotes neurotoxic amyloid β accumulation through reduced degradation by decreasing neprilysin protein levels in astrocytes.
Poly(I:C) promotes neurotoxic amyloid β accumulation through reduced degradation by decreasing neprilysin protein levels in astrocytes.
Journal of neurochemistry (2022-11-03)
Naoki Yamamoto, Takuya Tokumon, Ayako Obuchi, Mari Kono, Katsuyasu Saigo, Mamoru Tanida, Yuri Ikeda-Matsuo, Kazuya Sobue
ABSTRAKT
Inflammation associated with viral infection of the nervous system has been involved in the pathogenesis of neurodegenerative diseases, such as Alzheimer's disease (AD) and multiple sclerosis. Polyinosinic:polycytidylic acid (poly[I:C]) is a Toll-like receptor 3 (TLR3) agonist that mimics the inflammatory response to systemic viral infections. Despite growing recognition of the role of glial cells in AD pathology, their involvement in the accumulation and clearance of amyloid β (Aβ) in the brain of patients with AD is poorly understood. Neprilysin (NEP) and insulin-degrading enzyme (IDE) are the main Aβ-degrading enzymes in the brain. This study investigated whether poly(I:C) regulated Aβ degradation and neurotoxicity by modulating NEP and IDE protein levels through TLR3 in astrocytes. To this aim, primary rat primary astrocyte cultures were treated with poly(I:C) and inhibitors of the TLR3 signaling. Protein levels were assessed by Western blot. Aβ toxicity to primary neurons was measured by lactate dehydrogenase release. Poly(I:C) induced a significant decrease in NEP levels on the membrane of astrocytes as well as in the culture medium. The degradation of exogenous Aβ was markedly delayed in poly(I:C)-treated astrocytes. This delay significantly increased the neurotoxicity of exogenous Aβ1-42. Altogether, these results suggest that viral infections induce Aβ neurotoxicity by decreasing NEP levels in astrocytes and consequently preventing Aβ degradation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anty-IDE, N-końcowe (97-273) królicze pAb, liquid, Calbiochem®
Informacje o cenach i dostępności nie są obecnie dostępne.