Merck
Prulifloxacin.
Drugs (2004-10-01)
Susan J Keam, Caroline M Perry
ABSTRAKT
Prulifloxacin, the prodrug of ulifloxacin, is a broad-spectrum oral fluoroquinolone antibacterial agent. After absorption, prulifloxacin is metabolised by esterases to ulifloxacin. The drug has a long elimination half-life, allowing once-daily administration. Ulifloxacin is generally more active in vitro than other fluoroquinolones against a variety of clinical isolates of Gram-negative bacteria, including community and nosocomial isolates of Escherichia coli, Klebsiella spp., Proteus, Providencia and Morganella spp., Moraxella catarrhalis and Haemophilus spp. The activity of ulifloxacin against Pseudomonas aeruginosa varies between countries. Gram-positive organisms, including meticillin- or oxacillin-susceptible Staphylococcus aureus, Enterococcus spp. and Italian community isolates of Streptococcus pneumoniae are susceptible to ulifloxacin. Activity against Spanish strains of S. pneumoniae is moderate. In well designed clinical trials, good clinical and bacteriological efficacy (similar to that of ciprofloxacin, amoxicillin/clavulanic acid or pefloxacin) was seen with prulifloxacin 600 mg once daily for 10 days in patients with acute exacerbations of chronic bronchitis or complicated lower urinary tract infections (UTIs), and with single-dose prulifloxacin 600 mg in acute, uncomplicated lower UTIs. Prulifloxacin was generally well tolerated in clinical trials, with a similar tolerability profile to that of ciprofloxacin.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ulifloxacin, ≥98% (HPLC)
Informacje o cenach i dostępności nie są obecnie dostępne.