Merck
Targeting the CD40-CD40L pathway in autoimmune diseases: Humoral immunity and beyond.
Advanced drug delivery reviews (2018-12-16)
Jodi L Karnell, Sadiye Amcaoglu Rieder, Rachel Ettinger, Roland Kolbeck
ABSTRAKT
CD40 is a TNF receptor superfamily member expressed on both immune and non-immune cells. Interactions between B cell-expressed CD40 and its binding partner, CD40L, predominantly expressed on activated CD4+ T cells, play a critical role in promoting germinal center formation and the production of class-switched antibodies. Non-hematopoietic cells expressing CD40 can also engage CD40L and trigger a pro-inflammatory response. This article will highlight what is known about the biology of the CD40-CD40L axis in humans and describe the potential contribution of CD40 signaling on both hematopoietic and non-hematopoietic cells to autoimmune disease pathogenesis. Additionally, novel therapeutic approaches to target this pathway, currently being evaluated in clinical trials, are discussed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Protein delivery system, Human CD40, PODS®
Informacje o cenach i dostępności nie są obecnie dostępne.