Merck
Purification, crystallization and preliminary X-ray analysis of the membrane-bound [NiFe] hydrogenase from Allochromatium vinosum.
Acta crystallographica. Section F, Structural biology and crystallization communications (2008-08-06)
Petra Kellers, Hideaki Ogata, Wolfgang Lubitz
ABSTRAKT
The membrane-bound [NiFe] hydrogenase is a unique metalloprotein that is able to catalyze the reversible oxidation of hydrogen to protons and electrons during a complex reaction cycle. The [NiFe] hydrogenase was isolated from the photosynthetic purple sulfur bacterium Allochromatium vinosum and its crystallization and preliminary X-ray analysis are reported. It was crystallized by the hanging-drop vapour-diffusion method using sodium citrate and imidazole as crystallization agents. The crystals belong to space group P2(1)2(1)2, with unit-cell parameters a = 205.00, b = 217.42, c = 120.44 A. X-ray diffraction data have been collected to 2.5 A resolution.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Mineral oil, BioUltra, Molecular Biology
Sigma-Aldrich
Mineral oil, heavy
Sigma-Aldrich
Mineral oil, light oil, suitable for mouse embryo cell culture, BioXtra
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type II, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, suitable for protein sequencing, salt-free, lyophilized powder
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, (TLCK treated to inactivate residual tryspin activity), Type VII, essentially salt-free, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
Mineral oil, PCR Reagent
Sigma-Aldrich
Mineral oil, BioReagent, Molecular Biology, light oil
Sigma-Aldrich
Mineral oil, suitable for preparation of Nujol mulls for infrared spectroscopy, light oil
Sigma-Aldrich
Mineral oil, light oil, (neat)
Sigma-Aldrich
Mineral oil, meets USP testing specifications
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, ≥40 units/mg protein, vial of 5 mg
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type I-S, essentially salt-free, lyophilized powder
Sigma-Aldrich
Mineral oil, light