Merck
  • The human cancer antigen mesothelin is more efficiently presented to the mouse immune system when targeted to the DEC-205/CD205 receptor on dendritic cells.
The human cancer antigen mesothelin is more efficiently presented to the mouse immune system when targeted to the DEC-205/CD205 receptor on dendritic cells.
Annals of the New York Academy of Sciences (2009-09-23)
Bei Wang, Janelle M Y Kuroiwa, Li-Zhen He, Anna Charalambous, Tibor Keler, Ralph M Steinman
ABSTRAKT
To develop a tumor vaccine directly targeting tumor antigen to dendritic cells in situ, we engineered human mesothelin (MSLN) into an antibody specific for mouse DEC-205, a receptor for antigen presentation. We then characterized both T cell and humoral responses to human MSLN and compared immunizing efficacy of DEC-205-targeted MSLN to nontargeted protein after a single-dose immunization. Targeting human MSLN to DEC-205 receptor induced stronger CD4(+) T-cell responses compared to high doses of mesothelin protein. Approximately 0.5% CD4(+) T cells were primed to produce IFN-gamma, tumor necrosis factor-alpha, and IL-2 via intracellular cytokine staining, and the T cells also could proliferate rapidly. The immune response exhibited breadth because the primed CD4(+) T cells responded to at least three epitopes in the H-2(b) background. Targeting MSLN protein to DEC-205 receptor also resulted in cross-presentation to CD8(+) T cells. Antibody responses against human MSLN were also detected in serum from primed mice by ELISA assays. In summary, targeting of MSLN to DEC-205 improves the induction of CD4(+) and CD8(+) T-cell immunity accompanied by an antibody response. DEC-205-targeting could be valuable for enhancing immunity to MSLN in cancers where this nonmutated protein is expressed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Brefeldin A, Ready Made Solution, from Penicillium brefeldianum, 10 mg/mL in DMSO
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, Molecular Biology
Sigma-Aldrich
Monoclonal ANTI-FLAG® M1 antibody, clone M1, purified immunoglobulin, buffered aqueous solution