- Monoclonal antibody against human fibronectin which inhibits cell attachment.
Monoclonal antibody against human fibronectin which inhibits cell attachment.
Hybridoma (1982-01-01)
R C Schoen, K L Bentley, R J Klebe
PMID6208125
ABSTRAKT
Monoclonal antibodies have been prepared against both human and bovine fibronectin. Evidence is provided which indicates that nine different antigenic determinants are recognized by the ten antihuman fibronectin monoclonal antibodies isolated. One monoclonal antibody was identified that blocked fibronectin mediated cell attachment without interfering with fibronectin binding to collagen. Sensitive ELISA assays for fibronectins derived from 32 mammalian species have been developed with the monoclonal reagents characterized in this study.
MATERIAŁY
Numer produktu
Marka
Opis produktu
HFN 36.3, Reactivity: Anti-Fibronectin, 89062006, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Informacje o cenach i dostępności nie są obecnie dostępne.
HFN 7.1, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 89062001, Reactivity: Anti-Fibronectin, human
Informacje o cenach i dostępności nie są obecnie dostępne.