Merck
BI 2536, a potent and selective inhibitor of polo-like kinase 1, inhibits tumor growth in vivo.
Current biology : CB (2007-02-13)
Martin Steegmaier, Matthias Hoffmann, Anke Baum, Péter Lénárt, Mark Petronczki, Martin Krssák, Ulrich Gürtler, Pilar Garin-Chesa, Simone Lieb, Jens Quant, Matthias Grauert, Günther R Adolf, Norbert Kraut, Jan-Michael Peters, Wolfgang J Rettig
ABSTRAKT
Fine-mapping of the cell-division cycle, notably the identification of mitotic kinase signaling pathways, provides novel opportunities for cancer-drug discovery. As a key regulator of multiple steps during mitotic progression across eukaryotic species, the serine/threonine-specific Polo-like kinase 1 (Plk1) is highly expressed in malignant cells and serves as a negative prognostic marker in specific human cancer types . Here, we report the discovery of a potent small-molecule inhibitor of mammalian Plk1, BI 2536, which inhibits Plk1 enzyme activity at low nanomolar concentrations. The compound potently causes a mitotic arrest and induces apoptosis in human cancer cell lines of diverse tissue origin and oncogenome signature. BI 2536 inhibits growth of human tumor xenografts in nude mice and induces regression of large tumors with well-tolerated intravenous dose regimens. In treated tumors, cells arrest in prometaphase, accumulate phosphohistone H3, and contain aberrant mitotic spindles. This mitotic arrest is followed by a surge in apoptosis, detectable by immunohistochemistry and noninvasive optical and magnetic resonance imaging. For addressing the therapeutic potential of Plk1 inhibition, BI 2536 has progressed into clinical studies in patients with locally advanced or metastatic cancers.
MATERIAŁY
Numer produktu
Marka
Opis produktu
BI 2536, ≥98% (HPLC), powder, Plk1 inhibitor
Informacje o cenach i dostępności nie są obecnie dostępne.