Merck
Cell culture isolation of piscine neuropathy nodavirus from juvenile sea bass, Dicentrarchus labrax.
The Journal of general virology (1996-09-01)
G N Frerichs, H D Rodger, Z Peric
ABSTRAKT
A virus causing a vacuolating encephalopathy and retinopathy in juvenile sea bass, Dicentrarchus labrax, was isolated from brain tissue in a fish cell line (SSN-1) derived from striped snakehead, Channa striatus. The isometric, non-enveloped, 30 nm diameter virus particles were resistant to pH 2-9 and heating at 56 degrees C for 30 min. Infectious particles had a buoyant density of approximately 1.31 g/cm3 in CsCl. Two structural polypeptides of molecular mass 40 and 42 kDa were identified and the ssRNA consisted of two fragments of molecular mass 1.10 and 0.51 x 10(6) Da. From these characteristics the virus was identified as a nodavirus. Due to the broad range of susceptible fish hosts and the consistent neuropathology of the disease condition, the generic term piscine neuropathy nodavirus (PNN) is proposed for this infectious agent.
MATERIAŁY
Numer produktu
Marka
Opis produktu
SSN-1, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 96082808, fish unknown/unspecified, Fibroblast
Informacje o cenach i dostępności nie są obecnie dostępne.