Merck
Red fluorescent scaffold for highly sensitive protease activity probes.
Bioorganic & medicinal chemistry letters (2012-05-23)
Yu Kushida, Kenjiro Hanaoka, Toru Komatsu, Takuya Terai, Tasuku Ueno, Kengo Yoshida, Masanobu Uchiyama, Tetsuo Nagano
ABSTRAKT
We have developed a novel red fluorescent dye, 2Me SiR600 (λ(em)=613 nm), in which the O atom of Rhodamine Green at the 10 position of the xanthene moiety is replaced with a Si atom, as a scaffold for probes to detect protease activity with extremely high S/N ratio. As proof of concept, we designed and synthesized probes for caspase-3 activity (Z-DEVD-SiR600) and leucine aminopeptidase activity (Leu-SiR600). Caspase-3-mediated cleavage of Z-DEVD-SiR600 resulted in a large bathochromic shift (93 nm) of the absorption maximum and a 432-fold fluorescence enhancement.