Merck
Bioassay-guided isolation of urease and α-chymotrypsin inhibitory constituents from the stems of Lawsonia alba Lam. (Henna).
Fitoterapia (2012-10-30)
Nizam Uddin, Bina Shaheen Siddiqui, Sabira Begum, Muhammad Imran Ali, Bishnu P Marasini, Ajmal Khan, M Iqbal Choudhary
ABSTRAKT
Seven constituents were isolated from the stems of Lawsonia alba Lam., following an activity-guided isolation, which include two new constituents, namely lawsorosemarinol (1) and lawsofructose (2), one known compound 2-(β-d-glucopyranosyloxy)-1, 4-naphthoquinone (3) and four compounds, 4-hydroxy coumarine (4), 3-(4-hyroxyphenyl)-triacontyl-(Z)-propenoate (5), 3-(4-hydroxy-3-methoxyphenyl)-triacontyl-(Z)-propenoate (6) and 7-hydroxy-4-methyl coumarin (7) first time isolated from Lawsonia alba. Their structure elucidation was based on spectroscopic data analyses. Compounds 3 and 7 showed a moderate inhibition of urease activity, while rest of them showed less than 50% inhibition. These compounds did not show any significant inhibition against α-chymotrypsin.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type C-3, powder, ≥600,000 units/g solid
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type III, powder, 15,000-50,000 units/g solid
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean)
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type III, glycerol solution, 500-800 units/mL
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), powder, ~1 U/mg
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), powder, white, ~8 U/mg
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type IX, powder, 50,000-100,000 units/g solid