Merck
Acute effect of β-sitosterol on calcium uptake mediates anti-inflammatory effect in murine activated neutrophils.
The Journal of pharmacy and pharmacology (2012-12-12)
Rafael Liz, Leila Zanatta, Gustavo Oliveira dos Reis, Heros Horst, Moacir Geraldo Pizzolatti, Fátima Regina Mena Barreto Silva, Tânia Silvia Fröde
ABSTRAKT
To evaluate the effect of β-sitosterol on ⁴⁵Ca²⁺ uptake in activated murine neutrophils, and upon myeloperoxidase and adenosine deaminase activity, and interleukin-1β (IL-1β) and tumour necrosis factor-α (TNF-α) levels, in carrageenan-induced inflammation in the mouse air pouch model. Dried Esenbeckia leiocarpa bark was macerated and extracted resulting in a crude hydroalcoholic extract (CHE) that was partitioned to obtain an alkaloid fraction. The alkaloid was then partitioned in polar and nonpolar subfractions. β-Sitosterol was isolated from the nonpolar subfraction and identified by comparison with the literature. The effect of β-sitosterol on ⁴⁵Ca²⁺ uptake in activated murine neutrophils, and upon myeloperoxidase and adenosine deaminase activity, IL-1β and TNF-α levels in carrageenan-induced inflammation in mice were evaluated. β-Sitosterol promoted a time- and dose-dependent increase of the calcium uptake in activated neutrophils that was promptly reversed by nifedipine, BAPTA-AM, LY294002, and colchicine. β-Sitosterol inhibited myeloperoxidase and adenosine deaminase activity, and IL-1β and TNF-α levels. β-Sitosterol inhibited either myeloperoxidase and adenosine deaminase activity or IL-1β and TNF-α levels. This effect seemed to be mediated by the calcium uptake in activated neutrophils in a time- and concentration-dependent manner through L-type voltage dependent calcium channels, intracellular calcium, phosphoinositide kinase-3, and microtubule modulation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Adenosine Deaminase from bovine spleen, Type X, buffered aqueous glycerol solution, ≥130 units/mg protein
Sigma-Aldrich
Adenosine Deaminase from bovine spleen, Type IX, ammonium sulfate suspension, 150-200 units/mg protein
Sigma-Aldrich
β-Sitosterol, from soybean, ≥96%
Sigma-Aldrich
β-Sitosterol, synthetic, ≥95%
Sigma-Aldrich
β-Sitosterol, ≥70%