Merck
  • [The criteria of differentiated diagnostics of early arthritis on the basis of analysis of serum hyaluronidase and deoxyribonuclease activity].
[The criteria of differentiated diagnostics of early arthritis on the basis of analysis of serum hyaluronidase and deoxyribonuclease activity].
Klinicheskaia laboratornaia diagnostika (2012-12-26)
M V Volkova, E V Kunder
ABSTRAKT
The study analyzed serum hyaluronidase and deoxyribonuclease activity in patients with early arthritis--early rheumatoid arthritis and acute reactive arthritis. The criteria of their differential diagnostics were developed on the basis of data obtained. The genuine methods were applied to analyze hyaluronidase and deoxyribonuclease activity of blood serum based on formation of clot of etacridine acetate (rivanol) with hyaluronic acid and DNA inversely proportionally to their polymerization under the impact of enzymes. The increased serum hyaluronidase and deoxyribonuclease activity was established in patients with early arthritis as compared with control group (p < 0.001). The prevalence of mentioned types of activity under early rheumatoid arthritis as compared with acute reactive arthritis was detected too. The rests for differentiate diagnostics of early rheumatoid arthritis and acute reactive arthritis were developed conformed to criteria of the most useful diagnostic tests in rheumatology.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type VI-S, lyophilized powder, 3,000-15,000 units/mg solid
Sigma-Aldrich
Hyaluronidase from sheep testes, Type II, lyophilized powder, ≥300 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, powder, suitable for mouse embryo cell culture, 750-3000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, lyophilized powder (essentially salt-free), 750-3000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type VIII, lyophilized powder, 300-1,000 U/mg
Sigma-Aldrich
Hyaluronidase from sheep testes, Type V, lyophilized powder, ≥1,500 units/mg solid