Merck
Topoisomerase IIβ deficiency enhances camptothecin-induced apoptosis.
The Journal of biological chemistry (2013-01-25)
Ren-Kuo Lin, Chia-Wen Ho, Leroy F Liu, Yi Lisa Lyu
ABSTRAKT
Camptothecin (CPT), a topoisomerase (Top) I-targeting drug that stabilizes Top1-DNA covalent adducts, can induce S-phase-specific cytotoxicity due to the arrest of progressing replication forks. However, CPT-induced non-S-phase cytotoxicity is less well characterized. In this study, we have identified topoisomerase IIβ (Top2β) as a specific determinant for CPT sensitivity, but not for many other cytotoxic agents, in non-S-phase cells. First, quiescent mouse embryonic fibroblasts (MEFs) lacking Top2β were shown to be hypersensitive to CPT with prominent induction of apoptosis. Second, ICRF-187, a Top2 catalytic inhibitor known to deplete Top2β, specifically sensitized MEFs to CPT. To explore the molecular basis for CPT hypersensitivity in Top2β-deficient cells, we found that upon CPT exposure, the RNA polymerase II large subunit (RNAP LS) became progressively depleted, followed by recovery to nearly the original level in wild-type MEFs, whereas RNAP LS remained depleted without recovery in Top2β-deficient cells. Concomitant with the reduction of the RNAP LS level, the p53 protein level was greatly induced. Interestingly, RNAP LS depletion has been well documented to lead to p53-dependent apoptosis. Altogether, our findings support a model in which Top2β deficiency promotes CPT-induced apoptosis in quiescent non-S-phase cells, possibly due to RNAP LS depletion and p53 accumulation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DNA Gyrase from Escherichia coli, aqueous glycerol solution
Sigma-Aldrich
Dexrazoxane, ≥95% (HPLC)
Sigma-Aldrich
Przeciwciało anty-fosfoATM (Ser1981), klon 10H11.E12, clone 10H11.E12, Upstate®, from mouse
Sigma-Aldrich
Topoisomerase II α human, (Single band on SDS-PAGE), liquid
Sigma-Aldrich
Razoxane, >98% (HPLC)