- Selective oxidative decarboxylation of amino acids to produce industrially relevant nitriles by vanadium chloroperoxidase.
Selective oxidative decarboxylation of amino acids to produce industrially relevant nitriles by vanadium chloroperoxidase.
ChemSusChem (2012-05-05)
Andrada But, Jérôme Le Nôtre, Elinor L Scott, Ron Wever, Johan P M Sanders
PMID22556065
ABSTRAKT
Industrial nitriles from biomass: Vanadium-chloroperoxidase is successfully used to transform selectively glutamic acid into 3-cyanopropanoic acid, a key intermediate for the synthesis of bio-succinonitrile and bio-acrylonitrile, by using a catalytic amount of a halide salt. This clean oxidative decarboxylation can be applied to mixtures of amino acids obtained from plant waste streams, leading to easily separable nitriles.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Chloroperoxidase from Caldariomyces fumago, aqueous suspension, brown, >10,000 U/mL
Sigma-Aldrich
Chloroperoxidase from Caldariomyces fumago, buffered aqueous suspension, ≥3,000 units/mL
Sigma-Aldrich
Chloroperoxidase from Caldariomyces fumago, buffered aqueous suspension, 1,000-2,000 units/mg protein (E1%/280)