Merck
Synthesis, characterization, and bioavailability in rats of ferric phosphate nanoparticles.
The Journal of nutrition (2007-02-22)
Fabian Rohner, Frank O Ernst, Myrtha Arnold, Monika Hilbe, Ralf Biebinger, Frank Ehrensperger, Sotiris E Pratsinis, Wolfgang Langhans, Richard F Hurrell, Michael B Zimmermann
ABSTRAKT
Particle size is a determinant of iron (Fe) absorption from poorly soluble Fe compounds. Decreasing the particle size of metallic Fe and ferric pyrophosphate added to foods increases Fe absorption. The aim of this study was to develop and characterize nanoparticles of FePO(4) and determine their bioavailability and potential toxicity in rats. Amorphous FePO(4) nanopowders with spherical structure were synthesized by flame spray pyrolysis (FSP). The nanopowders were characterized and compared with commercially available FePO(4) and FeSO(4), including measurements of specific surface area (SSA), structure by transmission electron microscopy, in vitro solubility at pH 1 and 2, and relative bioavailability value (RBV) to FeSO(4) in rats using the hemoglobin repletion method. In the latter, the potential toxicity after Fe repletion was assessed by histological examination and measurement of thiobarbituric acid reactive substances (TBARS). The commercial FePO(4) and the 2 FePO(4) produced by FSP (mean particle sizes, 30.5 and 10.7 nm) had the following characteristics: SSA: 32.6, 68.6, 194.7 m(2)/g; in vitro solubility after 30 min at pH 1: 73, 79, and 85% of FeSO(4); and RBV: 61, 70, and 96%, respectively. In the histological examinations and TBARS analysis, there were no indications of toxicity. In conclusion, nanoparticles of FePO(4) have a solubility and RBV not significantly different from FeSO(4). Reducing poorly soluble Fe compounds to nanoscale may increase their value for human nutrition.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Iron(III) phosphate dihydrate, Fe 29 %
Informacje o cenach i dostępności nie są obecnie dostępne.