Merck
Biosynthesis, purification, and characterization of a cannabinoid receptor 2 fragment (CB2(271-326)).
Protein expression and purification (2008-04-01)
Yuxun Zhang, Xiang-Qun Xie
ABSTRAKT
Obtaining sufficient amount of purified G-protein coupled receptors (GPCRs) is almost always one of the major challenges for their structural studies. CB2(271-326), a human cannabinoid receptor 2 (CB2) fragment comprising part of the third extracellular loop (EL3), the seventh transmembrane domain (TM7) and C-terminal juxtamembrane region of the receptor, was over-expressed as a fusion protein into inclusion body (IB) of Escherichia coli. The fusion protein was purified by histidine-selected nickel affinity chromatography under denaturing conditions. Then, the fusion protein IBs were solubilized in detergent (Brij58) and the expression fusion leader sequence (TrpLE) was specifically cleaved with tobacco etch virus (TEV) protease. The target fragment, CB2(271-326), was subsequently purified by reverse-phase HPLC and confirmed by SDS-PAGE and mass spectrometry. This hydrophobic fragment can refold in mild detergents digitonin and Brij58. Circular dichroism (CD) spectroscopy of CB2(271-326) in digitonin and Brij58 micelles showed that the fragment adopts a more than 75% alpha-helical structure, with the remainder having beta-strand structure. Fluorescence spectroscopy and quenching studies suggested that the C-terminal region lies near the surface of the digitonin micelles and the TM7 region is folded relatively close to the center of the micelles. This study may provide an alternative strategy for the production and structure/functional studies of GPCRs such as CB2 receptor protein produced in the form of IBs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Brij® C10, average Mn ~683
Sigma-Aldrich
Brij® 58, average Mn ~1124
Sigma-Aldrich
SP Brij® C2 MBAL-SO-(SG), average Mn ~330
Sigma-Aldrich
ECO BRIJ® C10