Merck
  • Synthesis, characterization, luminescence, and non-linear optical properties of oxadiazole- and truxene-containing platinum(II) alkynyl complexes with donor-acceptor functionalities.
Synthesis, characterization, luminescence, and non-linear optical properties of oxadiazole- and truxene-containing platinum(II) alkynyl complexes with donor-acceptor functionalities.
Inorganic chemistry (2009-03-12)
Carmen Ka Man Chan, Chi-Hang Tao, Hoi-Lam Tam, Nianyong Zhu, Vivian Wing-Wah Yam, Kok-Wai Cheah
ABSTRAKT
A series of luminescent platinum(II) alkynyl complexes containing 2,5-diphenyl-1,3,4-oxadiazole (oxa-2,5) and 10,15-dihydro-5H-diindeno[1,2-a;1',2'-c]fluorene (truxene) as the cores and different electron donors at the periphery has been synthesized and characterized. These complexes showed long-lived emissions in both solution and in the solid state at room temperature and have been found to exhibit two-photon absorption (2PA) and two-photon induced luminescence. Their 2PA cross-sections (sigma(2)) have also been determined.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2,5-Diphenyloxazole, 99%, suitable for scintillation
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
2,5-Diphenyloxazole, suitable for liquid scintillation spectrometry
Informacje o cenach i dostępności nie są obecnie dostępne.