Merck
Highly efficient synthesis of DNA-binding hairpin polyamides via the use of a new triphosgene coupling strategy.
Organic letters (2009-08-13)
Wu Su, Stephen J Gray, Ruggero Dondi, Glenn A Burley
ABSTRAKT
A facile and highly efficient solid phase synthesis method is reported for the preparation of hairpin DNA-binding polyamides using the cost-effective triphosgene (BTC) activating agent. Difficult polyamide sequences were prepared from N-methylimidazole (Im) and N-methylpyrrole (Py) building blocks with high stepwise yields (>98%) using Boc chemistry. The versatility of the triphosgene coupling approach was also demonstrated for the first time on aryl hydrazine resins to afford biomedically relevant tail-truncated polyamides in excellent isolated yields.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
N-Methylpyrrole, 99%
Informacje o cenach i dostępności nie są obecnie dostępne.