Merck
Bioavailability and catabolism of green tea flavan-3-ols in humans.
Nutrition (Burbank, Los Angeles County, Calif.) (2010-01-19)
Daniele Del Rio, Luca Calani, Chiara Cordero, Sara Salvatore, Nicoletta Pellegrini, Furio Brighenti
ABSTRAKT
The aim of this study was to investigate green tea flavan-3-ol catabolism and plasma pharmacokinetic and urinary excretion by high-performance liquid chromatography with tandem mass spectrometry to evaluate their absolute bioavailability by taking into account all known and some unknown catabolites deriving from their interaction with the gastrointestinal tract and its host microflora. A feeding study was carried out in 20 healthy human volunteers who ingested 400 mL of a ready-to-drink green tea containing approximately 400 μmol of flavan-3-ols. Urine and plasma were collected for 4 and 24h, respectively, and 39 relevant catabolites were identified in these biological fluids by tandem mass spectrometry. In biological fluids, 39 relevant flavan-3-ol catabolites were identified. In plasma, (-)-epigallocatechin-3-gallate was the only unmetabolized compound and the highest in absolute concentration compared with (-)-epigallocatechin and (-)-epicatechin conjugates. Colonic microflora-derived polyhydroxyphenyl-γ-valerolactones were by far the main urinary catabolites, averaging 10 times greater concentratin than flavan-3-ol conjugates. The calculated bioavailability was equal to 39% and it is interesting to notice the great variability in urinary excretion of colonic metabolites among participants, probably related to differences in their own colonic microflora. This study demonstrates that green tea catechins are more bioavailable than previously observed when colonic ring fission metabolites are taken into consideration. Regular consumption of ready-to-drink green tea containing flavan-3-ols allows a non-marginal exposure of the human body to these catabolites, somehow justifying the numerous beneficial actions described as linked to green tea intake.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
(−)-Epicatechin, analytical standard
Sigma-Aldrich
4-Hydroxyphenylacetic acid, 98%
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
(−)-Epicatechin, ≥98% (HPLC), from green tea
Sigma-Aldrich
Pyrocatechol, ≥99%
Sigma-Aldrich
Homovanillic acid, Fluorimetric reagent
Sigma-Aldrich
(−)-Epicatechin, ≥90% (HPLC)
Epikatechina, primary reference standard
Supelco
4-Hydroxybenzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
γ-Valerolactone, ≥99%, FCC, FG
Sigma-Aldrich
1,2-dihydroksybenzen, ReagentPlus®, ≥99%
Sigma-Aldrich
γ-Valerolactone, ReagentPlus®, 99%
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
γ-Valerolactone, natural, ≥95%, FG
Supelco
Gallic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, ≥99%