Merck
Biodegradation of polyfluorinated biphenyl in bacteria.
Biodegradation (2010-09-11)
David Hughes, Benjamin R Clark, Cormac D Murphy
ABSTRAKT
Fluorinated aromatic compounds are significant environmental pollutants, and microorganisms play important roles in their biodegradation. The effect of fluorine substitution on the transformation of fluorobiphenyl in two bacteria was investigated. Pseudomonas pseudoalcaligenes KF707 and Burkholderia xenovorans LB400 used 2,3,4,5,6-pentafluorobiphenyl and 4,4'-difluorobiphenyl as sole sources of carbon and energy. The catabolism of the fluorinated compounds was examined by gas chromatography-mass spectrometry and fluorine-19 nuclear magnetic resonance spectroscopy (19F NMR), and revealed that the bacteria employed the upper pathway of biphenyl catabolism to degrade these xenobiotics. The novel fluorometabolites 3-pentafluorophenyl-cyclohexa-3,5-diene-1,2-diol and 3-pentafluorophenyl-benzene-1,2-diol were detected in the supernatants of biphenyl-grown resting cells incubated with 2,3,4,5,6-pentafluorobiphenyl, most likely as a consequence of the actions of BphA and BphB. 4-Fluorobenzoate was detected in cultures incubated with 4,4'-difluorobiphenyl and 19F NMR analysis of the supernatant from P. pseudoalcaligenes KF707 revealed the presence of additional water-soluble fluorometabolites.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Fluorobenzoic acid, 98%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
4-Fluorobenzoic acid, 99%, purified by sublimation
Informacje o cenach i dostępności nie są obecnie dostępne.