Merck
  • High-resolution structures of Trypanosoma brucei pteridine reductase ligand complexes inform on the placement of new molecular entities in the active site of a potential drug target.
High-resolution structures of Trypanosoma brucei pteridine reductase ligand complexes inform on the placement of new molecular entities in the active site of a potential drug target.
Acta crystallographica. Section D, Biological crystallography (2010-12-03)
Alice Dawson, Lindsay B Tulloch, Keri L Barrack, William N Hunter
ABSTRAKT
Pteridine reductase (PTR1) is a potential target for drug development against parasitic Trypanosoma and Leishmania species. These protozoa cause serious diseases for which current therapies are inadequate. High-resolution structures have been determined, using data between 1.6 and 1.1 Å resolution, of T. brucei PTR1 in complex with pemetrexed, trimetrexate, cyromazine and a 2,4-diaminopyrimidine derivative. The structures provide insight into the interactions formed by new molecular entities in the enzyme active site with ligands that represent lead compounds for structure-based inhibitor development and to support early-stage drug discovery.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Cyromazin, PESTANAL®, analytical standard
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
N-Cyclopropyl-2,4,6-triamino-1,3,5-triazine, 97%
Informacje o cenach i dostępności nie są obecnie dostępne.