Merck
Laser induced photocatalytic degradation of hazardous dye (Safranin-O) using self synthesized nanocrystalline WO3.
Journal of hazardous materials (2010-12-24)
K Hayat, M A Gondal, M M Khaled, Z H Yamani, S Ahmed
ABSTRAKT
The photocatalytic degradation of Safranin-O (known as Basic Red 2) in water using locally synthesized nanocrystalline WO(3) as a photocatalyst was investigated under UV laser irradiation. The photo-oxidation removal of the dye was monitored by UV-vis spectrophotometer. The blank experiments for either laser irradiated only Safranin-O solution or the suspension containing WO(3) and Safranin-O in the dark showed that both laser illumination and the photocatalyst were essential for the removal of Safranin-O. The effect of experimental parameters including laser energy, catalyst loading, solution pH and the initial dye concentration on photocatalytic degradation of Basic Red 2 were also investigated. Results indicate that the rate of reaction is strongly influenced by the adsorption of an azo dye into the surface of the photocatalyst materials and suggests an optimum catalyst loading and dye concentration for the degradation reaction. It was investigated that the adsorption of the dye decreases at higher alkaline pH because both catalyst and substrate are negatively charged, developing repulsive forces between them. Kinetic data obtained reveals that the rate of the reaction obeys the first-order kinetics.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Safranin O, Dye content ≥85 %
Sigma-Aldrich
Safranin O, suitable for microscopy
Sigma-Aldrich
Safranin O, certified by the BSC
Sigma-Aldrich
Schaeffer and Fulton Spore Stain Solution B
Sigma-Aldrich
Gram′s safranin solution, suitable for microscopy