Merck
Development of competitive immunoassays to hydroxyl containing fungicide metabolites.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2011-07-07)
Kevin C Gough, Shila Jarvis, Ben C Maddison
ABSTRAKT
This paper describes the isolation of monoclonal antibodies and the development of competitive immunoassays to pesticide metabolites of the fungicides imazalil, carbendazim and thiabendazole. The metabolite specific hydroxyl residues were used as the reactive group with which to link the metabolite to the carrier proteins Keyhole Limpet Haemocyanin (KLH) and Bovine Serum Albumin (BSA). In each case immune responses in mice were raised and monoclonal antibodies were produced. Antibodies were developed into competitive ELISAs to the appropriate metabolite. The antibody raised to a metabolite of imazalil was optimised into a competitive ELISA format which had an assay IC50 of 7.5 μg/L and a limit of detection (LOD) of 1.1 μg/L. A single antibody isolated against the metabolite of carbendazim had assay IC50s of 3.2 and 2.7 μg/L for the metabolites of carbendazim and thiabendazole respectively with an LOD of 0.38 μg/L for both. These sensitive immunoassays may have application in the monitoring of human exposure to these fungicide residues either by occupational or non-occupational routes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Thiabendazole, PESTANAL®, analytical standard
Sigma-Aldrich
Thiabendazole, BioReagent, suitable for plant cell culture, powder
Supelco
Thiabendazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Thiabendazole, ≥99%, powder
Supelco
Imazalil, PESTANAL®, analytical standard
Tiabendazole, European Pharmacopoeia (EP) Reference Standard