Merck
An unusual case of conjunctival irritation and epiphora following external dacryocystorhinostomy.
The Journal of laryngology and otology (2011-08-03)
P Mezzana, N Marabottini, F Scarinci, P Pasquini
ABSTRAKT
The authors discuss a case of conjunctival irritation and epiphora in a patient who had undergone dacryocystorhinostomy three years earlier. Case report, and a review of the world literature concerning lacrimal canalicular air regurgitation after dacryocystorhinostomy. A 70-year-old woman, who used continuous positive airway pressure therapy during sleep, complained of epiphora and conjunctival irritation. She had undergone dacryocystorhinostomy three years earlier. Her right eye had an augmented tear meniscus, with a predominantly medial conjunctival irritation. Conjunctival irritation from lacrimal canalicular air regurgitation was diagnosed. White petrolatum and mineral oil ophthalmic ointment was prescribed, applied topically before sleeping. After one month of treatment, complete relief of symptoms was obtained. There is little current information on managing such patients. It is imperative to discuss this potential complication when considering nasolacrimal surgery in patients using continuous positive airway pressure therapy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Petrolatum Jelly, meets analytical specification of Ph. Eur.
Informacje o cenach i dostępności nie są obecnie dostępne.