Merck
  • Preparation, characterization and application of a new stir bar sorptive extraction based on poly(vinylphthalimide-co-N,N'-methylenebisacrylamide) monolith.
Preparation, characterization and application of a new stir bar sorptive extraction based on poly(vinylphthalimide-co-N,N'-methylenebisacrylamide) monolith.
Journal of separation science (2011-11-05)
Xiaojia Huang, Linli Chen, Dongxing Yuan, Xianbin Luo
ABSTRAKT
In this study, a new stir bar sorptive extraction (SBSE) coating based on poly(vinylphthalimide-co-N,N'-methylenebisacrylamide) monolith (SBSE-VPMB) was prepared. The influences of the contents of monomer in polymerization mixture and the percentage of porogen solvent on the extraction performance were investigated thoroughly. Several characteristic techniques, such as elemental analysis, scanning electron microscopy, mercury intrusion porosimetry and infrared spectroscopy, were used to characterize the monolithic material. The analysis of oxfendazole (OFZ) and mebendazole (MBZ) in milk and honey samples by the combination of SBSE with HPLC with diode array detection was selected as paradigms for the practical evaluation of the new coating. Under the optimized extraction conditions, the limits of detection (S/N=3) for OFZ and MBZ were 0.23-0.60 μg/L in milk and 0.24-1.08 μg/L in honey, respectively. The method also showed good linearity, repeatability, high feasibility and acceptable recoveries for real samples. At the same time, the extraction performance and the distribution coefficients (K(VPMB/W)) of OFZ and MBZ on SBSE-VPMB were compared with other SBSEs based on porous monoliths and commercial SBSE.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
N,N′-Methylenebis(acrylamide), 99%
Sigma-Aldrich
N,N′-Methylenebisacrylamide, powder, Molecular Biology, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
N,N′-Methylenebisacrylamide, suitable for electrophoresis (after filtration or allowing insolubles to settle)
Sigma-Aldrich
N,N′-Methylenebisacrylamide solution, suitable for electrophoresis, 2% in H2O
Supelco
Oxfendazole, VETRANAL®, analytical standard