Merck
Alkylated dihydroxybenzoic acid as a MALDI matrix additive for hydrophobic peptide analysis.
Analytical chemistry (2012-04-18)
Yuko Fukuyama, Ritsuko Tanimura, Kazuki Maeda, Makoto Watanabe, Shin-Ichirou Kawabata, Shinichi Iwamoto, Shunsuke Izumi, Koichi Tanaka
ABSTRAKT
Hydrophobic peptides are generally difficult to detect using matrix-assisted laser desorption/ionization mass spectrometry (MALDI-MS) because the majority of MALDI matrixes are hydrophilic and therefore have a low affinity for hydrophobic peptides. Here, we report on a novel matrix additive, o-alkylated dihydroxybenzoic acid (ADHB), which is a 2,5-dihydroxybenzoic acid (DHB) derivative incorporating a hydrophobic alkyl chain on a hydroxyl group to improve its affinity for hydrophobic peptides, thereby improving MALDI-MS sensitivity. The addition of ADHB to the conventional matrix α-cyano-4-hydroxycinnamic acid (CHCA) improved the sensitivity of hydrophobic peptides 10- to 100-fold. The sequence coverage of phosphorylase b digest was increased using ADHB. MS imaging indicated that hydrophobic peptides were enriched in the rim of a matrix/analyte dried spot when ADHB was used. In conclusion, the addition of ADHB to the standard matrix led to improved sensitivity of hydrophobic peptides by MALDI-MS.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Gentisic acid sodium salt hydrate, ≥98%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 97%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, suitable for matrix substance for MALDI-MS, Ultra pure
Supelco
2,5-Dihydroxybenzoic acid, suitable for matrix substance for MALDI-MS, ≥99.5% (HPLC), Ultra pure
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, suitable for MALDI-TOF MS
Supelco
α-Cyano-4-hydroxycinnamic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Supelco
2,5-Dihydroxybenzoic acid, suitable for matrix substance for MALDI-MS, >99.0% (HPLC)
Sigma-Aldrich
2,5-Dihydroxybenzoic acid, 98%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, ≥98% (TLC), powder
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 99%