Merck
Tolfenamic acid induces apoptosis and growth inhibition in head and neck cancer: involvement of NAG-1 expression.
PloS one (2012-04-27)
Sung Un Kang, Yoo Seob Shin, Hye Sook Hwang, Seung Joon Baek, Seong-Ho Lee, Chul-Ho Kim
ABSTRAKT
Nonsteroidal anti-inflammatory drug-activated gene-1 (NAG-1) is induced by nonsteroidal anti-inflammatory drugs and possesses proapoptotic and antitumorigenic activities. Although tolfenamic acid (TA) induces apoptosis in head and neck cancer cells, the relationship between NAG-1 and TA has not been determined. This study investigated the induction of apoptosis in head and neck cancer cells treated by TA and the role of NAG-1 expression in this induction. TA reduced head and neck cancer cell viability in a dose-dependent manner and induced apoptosis. The induced apoptosis was coincident with the expression of NAG-1. Overexpression of NAG-1 enhanced the apoptotic effect of TA, whereas suppression of NAG-1 expression by small interfering RNA attenuated TA-induced apoptosis. TA significantly inhibited tumor formation as assessed by xenograft models, and this result accompanied the induction of apoptotic cells and NAG-1 expression in tumor tissue samples. Taken together, these results demonstrate that TA induces apoptosis via NAG-1 expression in head and neck squamous cell carcinoma, providing an additional mechanistic explanation for the apoptotic activity of TA.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Tolfenamic acid, VETRANAL®, analytical standard
Sigma-Aldrich
Tolfenamic acid, NSAID