Merck
The role of the IRE1 pathway in PBDE-47-induced toxicity in human neuroblastoma SH-SY5Y cells in vitro.
Toxicology letters (2012-05-01)
Chunyang Jiang, Shun Zhang, Hongliang Liu, Qiang Zeng, Tao Xia, Yihu Chen, Gang Kuang, Guodong Zhao, Xue Wu, Xiaofei Zhang, Aiguo Wang
ABSTRAKT
Polybrominated diphenyl ethers (PBDEs) are widely used as flame retardants. As one of the dominant congeners, 2,2', 4,4'-tetrabromodiphenyl ether (PBDE-47) has been shown to be neurotoxic to neuronal cells although the mechanisms remain unclear. To test whether PBDE-47's toxicity was related to endoplasmic reticulum (ER) stress and the unfolded protein response (UPR), human neuroblastoma cells (SH-SY5Y cells) were treated with different concentrations of PBDE-47. Reactive oxygen species (ROS), apoptosis and the expressions of the inositol-requiring enzyme 1 (IRE1) pathway-related molecules were detected. PBDE-47 exposure increased ROS production and activated the UPR by increasing the expressions of glucose-regulated protein 78 (GRP78), IRE1, X-box-binding protein-1 (XBP1), phosphorylation of c-jun N-terminal kinase (JNK) and GADD153/C/EBP homologous protein (CHOP) genes in SH-SY5Y cells. The apoptotic rate increased with the remarkable up-regulation of the Bax/Bcl-2 ratio after IRE1 knockdown, demonstrating the anti-apoptotic role of IRE1. Furthermore, the expressions of CHOP, XBP1 and JNK were down-regulated indicating that IRE1 may activate these key molecules related to apoptosis. PBDE-47 exposure can increase ROS production and activate the IRE1 pathway of the UPR in SH-SY5Y cells contributing to its toxicity. The IRE1 pathway may have both protective and proapoptotic effects on SH-SY5Y cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
bisBenzimide H 33258, suitable for fluorescence, ≥98.0% (HPLC)
Sigma-Aldrich
bisBenzimide H 33258, powder, BioReagent, suitable for cell culture, ≥98% (HPLC and TLC)
Sigma-Aldrich
bisBenzimide H 33258, ≥98% (HPLC and TLC)
Sigma-Aldrich
Hoechst 33258 solution, 1 mg/mL in H2O, ≥98.0% (HPLC)